The pump is shipped with a mixing system, comprising the following mixers:
Capillary mixer
In the capillary mixer, the solvent streams delivered by the pump are combined and premixed.
Static mixer
The static mixer improves the mixing quality.
Mixing with capillary mixer and static mixer results in smoother baselines.
The pump is shipped with the following mixing system:
Pump | Mixing System |
---|---|
Binary VC-pump Quaternary pump Dual pump | Mixing system, volume 400 µL, comprising:
The dual pump has two mixing systems. |
Binary VF-pump Isocratic pump | Mixing system, volume 200 µL, comprising:
With the isocratic pump, the mixing system serves as inline filter. |
For pump-specific pictures, see the Interior Components sections in Pump-Specific Information. |
TIP
Mixing systems with other volumes are available as an option (see Gradient Delay Volume, Dwell Volume and Ripple).