The purge/sensor unit comprises the purge valve and the system pressure sensor.
Purge/sensor unit (example, here in binary pump)
Figure: Purge/sensor unit (example, here in binary pump)
No.
Description
1a
Isocratic pump, dual pump, quaternary pump: Capillary from the pump head
Binary pump: Capillary from the right pump head
1b
Binary pump only: Capillary from the left pump head
2
System pressure sensor
3
Capillary mixer
4
Purge outlet
5
Purge valve knob
Binary Pump
With the binary pump, the purge/sensor unit also combines the solvent streams from the pump heads.