In the Chromeleon software, you can share the dual pump between two instruments.
The default assignment for the two pumps is the instrument in which the dual pump is installed.
To share the dual pump, follow these steps:
  1. In the Instrument Configuration Manager, open the dialog box for the dual pump.
  2. On the Devices tab page, assign each pump to the instrument on which you want to use the pump. Select the instrument from the list of available instruments.
TIP
Notice that the pumps now appear in the instruments to which they have been assigned.