The picture illustrates how the pump operates:
Operating principle
Figure: Operating principle
No.
Description
1
Solvent selector (pump inlet)
2
Solvent degasser (vacuum degasser)
3
Pump head, left, with
3a
Working cylinder (left pump head)
3b
Equilibration cylinder (left pump head)
4
Pump head right, with
4a
Working cylinder (right pump head)
4b
Equilibration cylinder (right pump head)
5
Purge/sensor unit, with
5a
Purge valve knob
5b
Purge outlet
5c
System pressure sensor
6
Mixing system, with
6a
Capillary mixer
6b
Static mixer
7
Pump outlet (outlet of the static mixer)
For additional information, see Operating Principle.