The user-accessible components of the pump are located directly behind the front doors:
Interior view
Figure: Interior view
No.
Description
1
Keypad with buttons for pump control
2a
Pump head (left)
2b
Pump head (right)
3
Tubing clips for routing the solvent lines from the degasser outlets to the pump heads
4
Purge/sensor unit including purge valve and system pressure sensor
5
Leak sensor
6
Drain port
7
Type label, indicating the module name, serial number, part number, and revision number (if any)
8
Seal wash detector (drop detector)
9a
Vacuum degasser (solvent B)
9b
Vacuum degasser (solvent A)
10a
Solvent selector (solvent B)
10b
Solvent selector (solvent A)
11
Seal wash pump
12
Tubing chase with tubing guides
13
Pump block LEDs
14a
Capillary mixer (as part of mixing system)
14b
Static mixer (as part of mixing system)
15
Pump outlet (outlet of the static mixer)