Note the following:
The dual pump combines two ternary pumps in a single enclosure.
The two pumps are referred to as left pump and right pump.
On the keypad, L refers to the left pump and R refers to the right pump.
In the Chromeleon user interface, Left refers to the left pump and Right refers to the right pump, for example, as in PumpLeft_Pressure and PumpRight_Pressure.
The dual pump can be shared by two different instruments (see Sharing the Dual Pump).
You can monitor the solvent consumption of the dual pump as a whole or separately for each pump (see Monitoring the Solvent Consumption).
You can monitor the liquid level in the waste container of the dual pump as a whole or separately for each pump (see Monitoring the Liquid Level in the Waste Container).