The picture illustrates how the pump operates:
Operating principle
Figure: Operating principle
No.
Description
A-D
Solvent options:
  • 4 solvents available with the quaternary pump (solvents A-D)
  • 3 solvents available with the dual pump (solvents A-C), with each of the two pumps
1
Solvent degasser (vacuum degasser) (pump inlet)
2
Proportioning valve
3
Pump head, with
3a
Working cylinder (pump head)
3b
Equilibration cylinder (pump head)
4
Purge/sensor unit, with
4a
Purge valve knob
4b
Purge outlet
4c
System pressure sensor
5
Mixing system, with
5a
Capillary mixer
5b
Static mixer
6
Pump outlet (outlet of the static mixer)
For additional information, see Operating Principle.