The user-accessible components of the pump are located directly behind the front doors:
Interior view
Figure: Interior view
No.
Description
1
Proportioning valve (3 channels) (left pump)
2
Tubing clips for routing solvent lines to the proportioning valve
3
Keypad with buttons for pump control
4
Pump head (left pump)
5
Purge/sensor unit including purge valve and system pressure sensor (left pump)
6
Proportioning valve (3 channels) (right pump)
7
Pump head (right pump)
8
Purge/sensor unit including purge valve and system pressure sensor (right pump)
9
Leak sensor
10
Drain port for guiding leaking liquid to waste
11
Type label, indicating the module name, serial number, part number, and revision number (if any)
12
Seal wash detector (drop detector)
13a
Vacuum degasser: Degas chambers for left pump, upper chambers (solvents AL BL CL)
13b
Vacuum degasser: Degas chambers for right pump, bottom chambers (solvents AR BR CR)
14
Seal wash pump
15
Tubing chase with tubing guides
16a
Capillary mixer (as part of mixing system, right pump)
16b
Static mixer (as part of mixing system, right pump)
17
Pump block LED (right pump)
18
Pump outlet (outlet of the static mixer) (right pump)
19a
Capillary mixer (as part of mixing system, left pump)
19b
Static mixer (as part of mixing system, left pump)
20
Pump block LED (left pump)
21
Pump outlet (outlet of the static mixer) (left pump)