The user-accessible components of the pump are located directly behind the front doors:
Interior view
Figure: Interior view
No.
Description
1
Keypad with buttons for pump control
2
Proportioning valve (4 channels)
3
Tubing clips for routing solvent lines to the proportioning valve
4
Pump head
5
Purge/sensor unit including purge valve and system pressure sensor
6
Leak sensor
7
Type label, indicating the module name, serial number, part number, and revision number (if any)
8
Drain port for guiding leaking liquid to waste
9
Seal wash detector (drop detector)
10
Vacuum degasser
11
Seal wash pump
12
Tubing chase with tubing guides
13
Pump block LED
14a
Capillary mixer (as part of mixing system)
14b
Static mixer (as part of mixing system)
15
Pump outlet (outlet of the static mixer)