The pump performance is specified as follows:
Type
Specification*
Operating principle
Serial dual-piston pump
Compressibility compensation
Fully automated, independent of the composition of the mobile phase
Flow range (settable)
VC-pumps:
0.001 – 10 mL/min in 1 µL/min increments
VF-pumps:
0.001 – 8 mL/min in 1 µL/min increments
Flow accuracy
± 0.1%
Flow precision
< 0.05% RSD or < 0.01 min SD, whichever is greater
Pressure range
VC-pumps:
2-70 MPa (20-700 bar, 290 – 10100 psi)
With a flow rate of > 5 mL/min, the maximum pressure decreases linearly down to 30 MPa (300 bar, 4350 psi).
VF-pumps:
2 – 103 MPa (20 – 1034 bar, 290 – 15000 psi)
With a flow rate of > 5 mL/min, the pressure range decreases linearly down to 80 MPa (800 bar, 11600 psi).
Pulsation
< 1.0% or < 0.2 MPa, whichever is greater
Gradient formation
Low-pressure gradient proportioning
Proportioning accuracy
± 0.5% (of full-scale)
Proportioning precision
< 0.15% SD
Number of solvents
VC-pumps:
VC-P20, VC-P21: 4
VC-P32, VC-P33: 2 x 3
VF-pumps:
VF-P20: 4
VF-P32: 2 x 3
Maximum stroke volume
80 µL
Mixer volume
Standard configuration: 400 µL mixing system, consisting of a capillary mixer (volume: 50 µL) and a static mixer (volume: 350 µL)
Other mixing systems are available as an option.
Dwell volume
(contribution of the pump to the system gradient delay volume)
679 µL (in standard pump configuration)
(314 µL to 1829 µL with optionally available mixing systems)
Solvent degassing
VC-pumps:
VC-P20, VC-P21:
Built‑in, 4 channels
VC-P32, VC-P33:
Built‑in, 6 channels
VF-pumps:
VF-P20:
Built-in, 4 channels
VF-P32:
Built-in, 6 channels
Biocompatible
VC-pumps: no
VF-pumps: yes
Communication
USB:
1 USB port (USB 2.0, "B" type connector)
1 USB hub with 3 ports (USB 2.0, "A" type connectors)
I/O Interface:
2 Dig I/O ports (mini-DIN), each providing one input, one relay output, and one bidirectional input/output
System Interlink:
2 system interlink ports (RJ45-8 connectors)
Control
Chromeleon 7
The pump can be operated also with other data systems. For details, contact the Thermo Fisher Scientific sales organization.
Keypad with 5 buttons for performing certain functions directly from the pump
Materials in the analytical flow path
VC-pumps:
Stainless steel, titanium, ceramics, sapphire, PEEK, UHMW PE, fluoropolymers
VF-pumps:
MP35N, titanium, ceramics, sapphire, PEEK, UHMW PE, fluoropolymers
For information about the chemical resistance of materials refer to the technical literature.
Solvent and additive information
See Solvent and Additive Information.
Safety features
Leak detection and safe leak handling, excess pressure monitoring
Good Laboratory Practice (GLP) features
Predictive Performance functions for scheduling maintenance procedures based on the actual operating and usage conditions of the pump.
All system parameters logged in the Chromeleon Audit Trail.
* Typical operating conditions for measurable specifications:
Flow accuracy, flow precision, pulsation: 1 mL/min at 20 MPa or 2 mL/min at 40 MPa, water
Proportioning accuracy, proportioning precision: 1 mL/min at 20 MPa or 2 mL/min at 40 MPa, 0-100%, water/spiked water