When you observe pressure issues or shifts in retention time, the following sections help you to identify and eliminate the source for the problem:
The pressure is not down to zero when the purge valve is open
Pressure pulsation
See Checking the Compression Values and Testing the Static Mixer for Permeability.
Shift in retention time
Inconstant pressure or high backpressure at the column and pump