When
Damage or blockage of the solvent line
Parts and additional items required
  • Solvent line between solvent reservoir and pump inlet
  • Solvent suitable for purging
Preparations
  1. To flush out harmful substances, purge the pump with an appropriate solvent.
  2. Empty the solvent lines (see Emptying the Solvent Lines).
Follow these steps
  1. Remove the solvent line from the reservoir cap:
  1. Remove the filter from the solvent line.
  2. Remove the retaining guide.
  3. Pull the solvent line out of the reservoir cap.
  1. Disconnect the solvent line on the pump inlet.
  2. Remove the solvent line from the pump and all modules above the pump in the system stack. When removing the solvent line from the tubing guides, do not pull on other tubing in the guides.
  3. To install the new solvent line, follow the related steps in Connecting the Solvent Lines.
  4. To remove any air from the solvent lines, purge the pump using the solvent of your application.