When
Damage of pump head
Parts required
  • Pump head
  • Spacing tool (pump head tool)
Tools and additional items required
  • Hexagon wrench, size 6
  • Depending on the capillary that connects the pump head to the purge/sensor unit:
    • Viper capillary: Installation tool for Viper capillaries with torque toothing
    • Conventional capillary (non-Viper): Open-end wrench, size 1/4"
  • Solvent suitable for purging to flush out harmful substances
  • Seal wash liquid
  • Solvent suitable for your application
Preparations
  1. To flush out harmful substances, purge the pump with an appropriate solvent.
  2. Turn off the pump flow and wait until the system pressure is down to zero.
Follow these steps
  1. Remove the pump head. See Removing the Pump Head.
  2. Install the pump head. See Installing the Pump Head.