The picture shows the main pump head parts.
Overview of pump head parts
Figure: Overview of pump head parts
No.
Description
1
Inlet check valve, including:
1a
Valve nut
1b
Valve cartridge
2
Pump head
3
Pump head seals (2 PTFE O-ring seals)
4
Piston seals
5
Support rings
6
Seal wash bodies (pump head bushing)
7
Seal wash seals
8
Seal wash plate
9
Pistons
10
Outlet check valve, including:
10a
Valve nut
10b
Valve cartridge
11
Connectors for seal wash tubing (pluggable)
12
U-tube (capillary from working cylinder to equilibration cylinder)