When
  • Contamination of detector electrodes
  • Impaired functionality of the seal wash system
Parts required
Seal wash detector assembly, consisting of the detector and funnel
Preparations
To prepare the new seal wash detector assembly push the seal wash detector into the funnel. Mind the orientation of the funnel.
1a1b2
Figure: Assembling the seal wash detector parts
No.
Description
1a
Funnel (bottom side)
1b
Funnel (top side)
2
Seal wash detector
Follow these steps
Removing the seal wash detector assembly
Figure: Removing the seal wash detector assembly
No.
Description
1
Funnel, seal wash detector
2
Seal wash detector
3
Detector cable
4
Seal wash line
  1. Disconnect the seal wash line from the seal wash detector (port labeled in).
  2. Unplug the detector cable from the DROP DET port.
  3. Grasp and pull the seal wash detector assembly upward by the funnel.
  4. Push the new seal wash detector assembly into the pump.
  5. Figure: Installing the seal wash detector assembly
  6. Reconnect the seal wash line to the seal wash detector (port labeled in).
  7. Connect the detector cable to the DROP DET port.
  1. Flush the seal wash system, using fresh seal wash liquid (see Flushing the Seal Wash System).