Each mixing system comprises a static mixer and a capillary mixer. The volumes of both mixers determine the entire volume of the mixing system.
The table lists the available mixing systems:
Description
Remarks
Set inline filter, volume: 35 µL, comprising:
  • Inline filter (static filter), volume: 10 µL
  • Capillary mixer, volume: 25 µL
Requires also a different capillary to connect the pump to the autosampler.
Mixing system, volume: 100 µL, comprising:
  • Static mixer, volume: 75 µL
  • Capillary mixer, volume: 25 µL
Requires also a different capillary to connect the pump to the autosampler.
Mixing system, volume: 200 µL, comprising:
  • Static mixer, volume: 150 µL
  • Capillary mixer, volume: 50 µL
Binary VF-pump, isocratic pump: Mixing system with which the pump is shipped.
With the isocratic pump, the mixing system serves as inline filter.
Other pumps: See TIP at the end of this table.
Mixing system, volume: 400 µL, comprising:
  • Static mixer, volume: 350 µL
  • Capillary mixer, volume: 50 µL
Binary VC-pump, quaternary pump, dual pump: Mixing system with which the pump is shipped.
The dual pump has two mixing systems.
Other pumps: See TIP at the end of this table.
Mixing system, volume: 800 µL, comprising:
  • Static mixer, volume: 750 µL
  • Capillary mixer, volume: 50 µL
See TIP at the end of this table.
Mixing system, volume: 1550 µL, comprising:
  • Static mixer, volume: 1500 µL
  • Capillary mixer, volume: 50 µL
See TIP at the end of this table.
TIP With this mixing system, only the static mixer is different from the static mixer with which the pump is shipped. The capillary mixer remains the same. Thus, if you want to change to this mixing system, you need to order only the static mixer.
For ordering information, see Optional Accessories.
To change to a different mixing system, see Exchanging the Mixing System.