Parts required
Capillary to connect the pump to the autosampler, as required for the flow path
Follow these steps
  1. On the connecting capillary, remove the knurls.
  2. Guide the connecting capillary through the guide holes in the housings of both, the pump and autosampler.
  3. Guide holes in the autosampler and pump (left: view from VF‑/VH-autosampler (example); right: view from pump)
    Figure: Guide holes in the autosampler and pump (left: view from VF‑/VH-autosampler (example); right: view from pump)
    No.
    Description
    1
    Capillary connecting the pump and autosampler:
    1a
    Capillary end connected to the autosampler injection valve
    1b
    Capillary end to be connected to pump outlet
    2
    Guide holes in autosampler housing and pump housing:
    2a
    Guide hole in autosampler housing (view from above)
    2b
    Guide hole in pump housing (view from below)
  4. Reattach the knurls to the connection capillary.
  5. In the pump, connect the capillary to the pump outlet.
  6. In the autosampler, connect the capillary to the injection valve.