The following liquids collect in the leak tray and flow off through the drain port at the bottom right of the leak tray into the drain system of the Vanquish system:
  • Leaking liquid from the leak tray
  • Waste liquid from the purge unit: When purging the pump, attach silicone tubing to the purge outlet on the purge/sensor unit and guide the free end of the tubing to the drain port outlet.
Drain port (view from top)
Figure: Drain port (view from top)
No.
Description
1
Waste line from purge unit: routing path to drain port outlet (example)
2
Drain port
3
Drain port outlet (to Vanquish drain system)
4
Leak sensor
For information about how the liquid is discharged to waste through the Vanquish drain system, refer to the Vanquish System Operating Manual.