Automatic testing means that the test runs automatically without further user interaction. The test checks the tightness of the pump and autosampler.
Prerequisite
A compatible chromatography data system that supports basic tightness testing for the configuration of your Vanquish system
Follow these steps
In the chromatography data system, run the Basic Tightness Test from the Diagnostic Tests menu.
If the test fails, check the Instrument Audit Trail for a message.
Take remedial action as suggested for the message in the Troubleshooting section of Operating Manual for the module for which the test failed.