For successful maintenance and service procedures, follow the rules and recommendations below.
General Rules
  • Before starting maintenance or service procedures, shut down the device when instructed to do so.
  • Use only the replacement parts specifically authorized and qualified for the device by Thermo Fisher Scientific.
  • Follow all instructions step by step and use the tools recommended for the procedure.
Opening Flow Path Connections
  • Before opening the flow path to replace capillaries in the system, turn off the pump flow and wait until the system pressure is down to zero.
  • Dirty components can contaminate the chromatography system. Contamination leads to poor performance of the modules and entire system or can even cause damage to the modules and system. Therefore:
    • Always wear appropriate gloves.
    • Place the components only on a clean, lint-free surface.
    • Keep your tools clean.
    • Use only lint-free cloth for cleaning.
  • After you have performed maintenance or service on the flow connections in the pump, test the pump for leakage before you resume operation.
Depot Repair