Device Connectors
The following connectors are provided on the device:
Electrical connectors and device connectors on the pump
Figure: Electrical connectors and device connectors on the pump
No.
Description
1
Rating plate (example), indicating the serial number, part number, module name, revision number (if any), and the line and fuse rating
2
Main power switch (on/off control)
3
Fuse holder
4
Power-inlet connector
5
System Interlink port
Allows power on/off control for the device from the Vanquish system base and device communication
6
Digital I/O ports (Dig I/O)
Allow exchange of digital signals with external instruments
Each digital I/O port provides one input, one relay output, and one bidirectional input/output. For connection and pin assignment information, see Digital I/O.
7
USB hub ("A" type connector)
Allows connection to other modules in the Vanquish system
8
USB (Universal Serial Bus) port ("B" type connector)
Allows connection to other modules in the Vanquish system or the computer on which the data management system is installed, such as the Chromeleon software
TIP
Thermo Fisher Scientific recommends using the USB ports only as described above. If the USB ports are used for any other purpose, Thermo Fisher Scientific cannot ensure proper functionality.
Follow these steps
NOTICE
  • Never use defective communication cables. If you suspect that a cable is defective, replace the cable.
  • To ensure trouble-free operation, use only the cables provided by Thermo Fisher Scientific for connecting the device.
  1. Place the device in the system as required by the system configuration. For details, refer to the Vanquish System Operating Manual.
  2. Connect the required interface cables to the device. For information about how to connect the device to other modules in the Vanquish system or to the chromatography data system computer, refer to the Vanquish System Operating Manual.
  3. Connect the power cord (see Connecting the Power Cord).