When operating the device, pay attention to the following safety guidelines:
Observe all warning messages and precautionary statements presented in Safety Precautions.
CAUTION
High Luminosity of LED
The high luminosity produced by the LED illuminating the inside of the device can be harmful to the eyes.
  • Do not look directly into the light produced by the LED.
  • Do not use light-focusing instruments for viewing the light beam.
NOTICE
Pay attention also to the following guidelines:
  • To prevent damage resulting from leakage or from running the pump dry, always set the lower pressure limit for the pump.
  • If there is evidence of leakage in the device, turn off the pump flow and remedy the situation immediately.
  • If the pump flow is interrupted, act appropriately to protect the components in the detector. For details, refer to the Operating Manual for the detector.
  • Always verify that the autosampler is turned on before the pump flow is on and pressure builds up. If the autosampler is turned off, for example, after a power failure, stop the pump flow and wait until the pressure is down to zero before turning on the autosampler or other modules again.