The digital I/O ports (Dig I/O) can be used to exchange digital signals with external devices. Each port provides:
  • one digital input
  • one relay output
  • one bidirectional input/output
Pin Assignment
Digital I/O port
Figure: Digital I/O port
Pin
Description — Signal Name
1
Bidirectional input/output
2
Relay output — Relay_NC (Normally Closed contact)
3
Ground — GND
4
Digital input — Input
5
Relay output — Relay_COM
COM is the common contact for NO and NC. If the relay is not activated or if the device is turned off, the connection is between COM and NC. If the relay is activated, the connection is between COM and NO.
6
Relay output — Relay_NO (Normally Open contact)
The next table lists the functions assigned to the connector pins and the color of the cable wire connected to each pin.
Pin
Wire Color
Signal Name
Signal Level
Remarks
1
Pink
Input/output
Input (low active):
On: 0-0.4 V
Off: 2.2-5 V
Open Collector Output:
0-5 V, 0-2 mA
Pull-up resistor: 47 kΩ to 5 V
Configure as either input or output. Reference potential is ground. Note the following:
  • The maximum input voltage at the input must not exceed +5 V with reference to ground.
  • The minimum input voltage must not be lower than the ground potential.
2
Gray
Relay output — Relay_NC
Potential free
0-24 V, 0-100 mA
Opening contact
3
Green
Ground — GND
Ground
Reference potential
4
Yellow
Digital input — Input
Input (low active):
On: 0-0.4 V
Off: 2.2-5 V
Pull-up resistor: 47 kΩ to 5 V
Digital input; reference potential is ground. Note the following:
  • The maximum input voltage at the input must not exceed +5 V with reference to ground.
  • The minimum input voltage must not be lower than the ground potential.
5
White
Relay output — Relay_COM
Potential free
Common contact for NO and NC
6
Brown
Relay output — Relay_NO
Potential free
0-24 V, 0-100 mA
Closing contact
Prerequisites
To use the digital I/O functionality, the following must be fulfilled:
  • The digital I/O port is connected to the external device with the digital I/O signaling cable.
  • The inputs and outputs that you want to use are selected in the Instrument Configuration Manager.
Connecting a Digital I/O Port
  1. Plug the 6-pin connector of the cable into the digital I/O port that you want to use.
  2. For each relay output or digital input to be used, connect the appropriate signal wire and ground wire to the corresponding connectors on the external device. For details, refer to the documentation provided with the external device.
Selecting the inputs and outputs in the chromatography data system
  1. In the dialog box for the pump, on the Inputs and Outputs pages, select the inputs (Pump_Input_X) and outputs (Pump_Relay_X) that you want to use. The numbering in the dialog box corresponds to the port numbers on the pump.
  2. To configure the bidirectional input/output (Pump_IO_X), select the related check box on either the Inputs or Outputs page, depending on the preferred usage.