To prepare the device for transport, follow these steps:
  1. Perform a long-term shut down of the device (see Long-Term Shutdown).
  2. Turn off the device with its main power switch and disconnect the power cord.
  3. Remove all cables and flow connections to other devices.
  1. Close open flow connections with appropriate plugs, for example, the plugs you removed when setting up the pump.
  2. Check the leak tray and drain port. If liquid is present, absorb the liquid with a tissue.
    1. Remove the device or slide-in module from the system stack as required:
      Install the carrying handles and remove the device from the Vanquish system. Follow the instructions on dismounting the system stack in the Transporting or Shipping the System section of the Vanquish System Operating Manual.
      ‑or‑
      Remove the slide-in module from the device enclosure in the system stack (see Removing the Slide-In Module).