The device is designed to be operated mainly from a computer running with the chromatography software.
In addition, the following elements are available on the device:
  • Keypad
    The keypad buttons allow you to perform certain functions directly from the device.
  • Status indicators
    The LEDs (Light Emitting Diodes) on the status indicator LED bar on the front side of the device and the STATUS LED on the keypad provide a quick visual check of the operational status of the device.
Pump Block Status LED
The status indicator LED is located inside the pump above the pump head (pump block LED). The LED indicates the operational status of the pump block.