NOTICE
Condensation in a device can damage the electronics.
Verify that the power switch on the device is set to OFF.
Connect the power cord to the power inlet connector on the device.
Connect the free end of the power cord to an appropriate power source.