Damaged Packaging, Defective on Arrival
Inspect the shipping container for signs of external damage and, after unpacking, inspect the device for any signs of mechanical damage that might have occurred during shipment.
If you suspect that the device may have been damaged during shipment, immediately notify the incoming carrier and Thermo Fisher Scientific about the damage. Shipping insurance will compensate for the damage only if reported immediately.
Unpacking the Device
CAUTION
Heavy Load, Bulky Device
The device is too heavy or bulky for one person alone to handle safely. To avoid personal injury or damage to the device, observe the following guidelines:
  • Physical handling of the device, including lifting or moving, requires a team effort of two persons.
  • A team effort is in particular required when lifting the device into the system stack or when removing it.
  • Use the carrying handles that were shipped with the device to move or transport the device. Never move or lift the device by the front doors. This will damage the doors or the device.
Tools required
Screwdriver, TorxTM T20
Follow these steps
  1. Place the shipping container on the floor and open it.
  2. Remove the ship kit from the shipping container.
  3. Remove the device from the shipping container: Grasp the device by the carrying handles. Slowly and carefully, lift the device out of the shipping container.
  4. Carrying handles on the device
    Figure: Carrying handles on the device
    No.
    Component
    1
    Carrying handles
    2
    Attachment screw (one on each carrying handle)
  5. Place the device on a stable surface.
  6. If applicable:
    Remove any additional packing material. Leave any protective films attached to the surfaces of the device until it is properly positioned in the system stack.
  7. Transport the device by the carrying handles to the installation site, if it is not already there, and place it in the system stack (see System Arrangement).
  8. On each carrying handle, loosen the attachment screw until the carrying handle is moveable in the rail. Do not remove the screws from the carrying handles completely.
  9. Slide off the carrying handles from the rails towards the rear of the device.
  10. Sliding off the carrying handle from the left rail
    Figure: Sliding off the carrying handle from the left rail
  11. TIP
    Keep the shipping container, the carrying handles with the attachment screws, and all packing material. These items will be needed if the device is transported to a new location or shipped.
  12. Some surfaces including the doors of the device are covered by a protective film during shipment. Remove the protective film from all surfaces as applicable.