321654
Figure: Metering device head overview
No.
Description
1
Metering device head
2
Attachment screws
3
Seal wash outlet
4
Capillary to pressure sensor
5
Capillary to injection valve
6
Seal wash inlet