The following icons are present in the navigation bar.
Icon | Description |
|---|---|
Home panel Gives access to the functional parameters of all system modules attached to the system controller via the system interlink interface. For further information, see Home Panel. | |
Script panel Provides automated scripts. For further information, see Script Panel. | |
Notification panel Stores information about all warnings and errors of the system modules and all events of the system controller since the last system controller reboot For further information, see Messages. | |
Settings panel For further information, see Settings Panel. | |
Information panel Shows various information, for example, attached system modules and their serial numbers | |
Online plot panel (available on the home panel) Opens a dialogue box showing the connected modules on the top half and the associated physical value in the bottom half. The plot is updated continuously and shows the signal of the last 10 minutes. | |
Filter (only available on the script panel and the notification panel) Lets you filter by a system module, for example. |