This section provides information on messages that the system modules have in common.
The table lists the most frequently observed messages for the system modules.
Message and Code | Description and Remedial Action |
---|---|
|
xx = two-digit to four-digit code number When the message appears, write down the message code and turn off the module. Wait for 5 seconds and turn on the module again. If the message appears again, contact Technical Support. |
|
xx = two-digit to four-digit code number When the message appears, write down the message code. Turn off the module and contact Technical Support. |
|
xx = the number of seconds within the leakage must be resolved before the sequence will be aborted Find and eliminate the source for the leakage. If leakage in the pump is not resolved after 3 minutes, the pump stops the flow. 1. Visually inspect all components and connections in the flow path. 2. Tighten or replace the connection or component as required. 3. With a cloth or tissue, absorb all liquid in the leak tray and under the leak sensor. |
|
The message appears because the leakage reported earlier with code 33 was not resolved within 3 minutes. The pump stopped and/or the running sequence was aborted because of leakage. Find and eliminate the source for the leakage (see Testing for Liquid Leaks). |
|
The firmware download has not been successful. Repeat the download. |
|
The firmware download has not been successful. Verify that the correct firmware file was selected. Repeat the download. |
|
The firmware is incomplete, for example, because the communication between the chromatography data system and the module was interrupted during the firmware download. Repeat the download. |
|
The message may appear for a pressure sensor in a pump head or for the system pressure sensor in the flowmeter (see identification information in Pump Messages).
|
|
X = left valve or right valve The respective switching valve may be defective. Replace the switching valve, see Replacing the Vanquish Switching Valve or Replacing the VICI Switching Valve. If the error message persists, contact Technical Support. |
|
Contact Thermo Fisher Scientific Technical Support for assistance. Note that the module can be operated nevertheless, even if the leak sensor is defective or not present, if leak detection is disabled. |
|
You tried to download an incompatible firmware with an earlier version number than the firmware that is currently installed in the module. Downgrading the firmware may result in loss of functionality or malfunctioning of the module. If required, repeat the download with a firmware version later than the version currently installed in the module. |
|
All modules:
In addition for the pump: If the message appears for a pressure sensor in a pump head or for the system pressure sensor in the flowmeter (see identification information in Pump Messages):
|
|
All modules:
In addition for the pump: If the message appears for a pressure sensor in a pump head or for the system pressure sensor in the flowmeter (see identification information in Pump Messages):
|
| This is a software problem. The module produces data faster than the computer on which the chromatography data system is running can process the data. 1. In the chromatography data system, disconnect and reconnect the module. 2. If this does not solve the problem, update the firmware or the chromatography data system version. 3. If the problem persists: Also, third-party software on the computer, for example, virus scanners or poor computer performance can cause the problem. Contact the onsite IT department. |
|
Communication with the module failed. The module did not respond in time. For the module for which the message appears: 1. Turn on the module if it is not yet turned on. 2. Check the system interlink connections to the module. Verify that all system interlink cables are connected at both ends. 3. If the message persists, replace the system interlink cables. |
| All other modules on the system interlink bus are turned off or a firmware download is performed on all other modules. 1. Check whether all other modules have been turned off or a firmware download is performed on all other modules on the bus. 2. Wait until one of the modules on the bus is turned on or the firmware download is complete. 3. A system interlink cable may be disconnected from the module or is defective. Check the system interlink connections to the module. Verify that all system interlink cables are connected at both ends. 4. If the message persists, replace the system interlink cables. |
|
A system interlink cable may be disconnected from the module or defective.
|
|
X = serial number The module with the given serial number may be disconnected, turned off or a firmware update is running.
|
|
A system interlink cable may be disconnected from the module or defective. 1. Check the system interlink connections to the module. Verify that all system interlink cables are connected at both ends. 2. If the message persists, replace the system interlink cables. |
| Control access is not allowed via the system controller. You can only modify the module settings via the chromatography data system. Wait until, for example, the sequence has finished and the LED bar is green. Then repeat the action via the system controller. |
| Control access is not allowed via the chromatography data system. You can only modify the module settings via the system controller. Wait until, for example, the script has finished and the LED bar is green. Then repeat the action via the chromatography data system. |
| X = lock holder ID, with keypad button ID, USB address referring to the chromatography data system or system interlink address referring to the system controller or a module The module is already locked by another software (system controller or chromatography data system) or a keypad button. Wait until the module is released from the locked state. |
| X = lock holder ID, with keypad button ID, USB address referring to the chromatography data system or system interlink address referring to the system controller or a module Inform Thermo Fisher Scientific about the occurrence. No further action required. |
| X = lock holder ID, with keypad button ID, USB address referring to the chromatography data system or system interlink address referring to the system controller or a module The lock was released automatically since its holder disappeared from system interlink. 1. Check if the module holding the lock was turned off. 2. Check the system interlink connections to the module. |
|
Module is not ready to accept command or property assignment since locked by another software (system controller or chromatography data system). Wait until module is released by the current software (system controller or chromatography data system). |