When
Sample loop is clogged or damaged
Different sample loop volume needed
Parts required
Replacement sample loop or sample loop with different size
Follow these steps
Remove the sample loop.
Install the replacement sample loop.
Complete the replacement procedure and choose the corresponding sample loop in the chromatography data system.
TIP
It is also a script available that guides you through the sample loop replacement. If you need guidance, run the script: A07 - Change Sample Loop.