The user-accessible components are located directly behind the front doors.
2133c3b3a3d3e3f45768
Figure: VN-A10 Autosampler interior view
No.
Description
1
Sample compartment with carousel
2
Drain pump
3
Leak tray with:
3a
Keypad with status indicators
3b
Pump capillary guide hole
3c
Type label, indicating the module name, serial number, part number and revision number
3d
Drain funnel
3e
Drain ports
3f
Leak sensor
4
Needle wash pump
5
Injection unit (refer to next page for detailed view)
6
Insulation Cover
7
Seal wash inlet port
8
Guides for capillary to column compartment
Injection Unit of the Autosampler
12395678410
Figure: VN-A10 Detailed view of the injection unit without insulation cover
No.
Description
1
Shipping lock screw for needle unit
2
Needle unit with vial pusher
3
Sample loop unit
4
Needle seat
5
Injection valve
6
Solvent valve
7
Check valve
8
Selector valve
9
Metering device head
10
Pressure sensor