This section provides information on messages with code numbers 2000 through 2999. These messages are specific to the column compartment.
The table lists the most frequently observed messages for the column compartment.
Message and Code | Description and Remedial Action |
---|---|
|
xx = two-digit to four-digit code number When the message appears, write down the message code and turn off the module. Wait for 5 seconds and turn on the module again. If the message appears again, contact Technical Support. |
|
xx = two-digit to four-digit code number When the message appears, write down the message code. Turn off the module and contact Technical Support. |
|
Verify that the configured valve pod and the installed valve pod match. |
|
Verify that the configured valve pod and the installed valve pod match. |