The drain pump (peristaltic pump) removes any condensation from the sample compartment that may occur during sample cooling and drains it through a separate drain port to waste.
123456
Figure: Drain pump
No.
Description
1
Pump lever
2
Drain pump rotor
3
Peristaltic tubing
4
Tubing holder
5
Condensation tubing to sample compartment
6
Waste tubing to drain port