CAUTION
Heavy Load, Bulky Device
The system modules are too heavy or bulky for one person alone to handle safely. To avoid personal injury or damage to the system modules, observe the following guidelines:
  • Physical handling of the modules, including lifting or moving, requires a team effort of two persons.
  • A team effort is in particular required when lifting the modules into the system stack or when removing them.
Tools required
Screwdriver, Torx T20
Preparations
See Preparing the Autosampler for Removal.
Removing the Slide-in Module
  1. Loosen the four captive screws on the front left and front right of the device.
  2. Figure: Captive screws on the autosampler slide-in module
  3. Push all tubing and capillaries, which are present in the tubing chase of the Vanquish system modules, into the tubing chase.
  4. Grasp the slide-in module by the middle frame where the door lock is placed, and pull the module out of the enclosure by approximately 10 cm.
NOTICE
The slide-in module can fall down when pulling it out of the enclosure too far.
Pull out the slide-in module just far enough so that you can grasp it on both sides from below.
  1. Remove the slide-in module from the enclosure. The following steps require a team effort:
  1. Take the slide-in module on both sides from below.
  2. Pull the slide-in module from the rails towards the front.
  3. Place the slide-in module on a clean and stable surface.