The system performance is specified as follows:
Type | Specification |
|---|---|
Communication | The system base provides the following ports: 1 System Interlink port (RJ45‑8 connector) 1 VSC port (RJ-12 connector) For ports on the system modules, see the performance specification sections for the system modules. |
Control | Vanquish User Interface Keypad with buttons for performing certain functions directly from the system module. The system can be operated with a chromatography data system. For details, contact the Thermo Fisher Scientific sales organization. |
Materials in the flow path | For materials in the flow path, refer to the performance specification sections for the pump, autosampler, and the column compartment. |
Solvent and additive information | |
Biocompatibility | Yes |
Safety features | Leak detection and safe leak handling, excessive pressure monitoring |
Good Laboratory Practice (GLP) features | Predictive Performance functions for scheduling maintenance procedures based on the actual operating and usage conditions of the system modules. All system parameters logged on the notification panel. |