Available Power Buttons and Main Power Switches
The image shows the powering devices of the system.
No. | Description |
|---|---|
1 | System power button |
2 | System Controller power button |
3 | Main power switches |
System Power Button
If the main power switches of the system modules are on, you can use the system power button for turning on/off all system modules (except the system controller) simultaneously to facilitate the turn on/off control. Observe the following:
Status | Description |
|---|---|
On |
|
Off |
|
System Controller Power Button
The system controller power button is a push button and is completely independent of the system power button. If the system controller is turned on, the display is automatically turned on. After a power outage, the system controller restarts automatically.
To turn off the system controller and the display (if available), use the system controller power button and wait until the status LED (Light Emitting Diode) is off.