The cable clips on the system's rear can be used to guide the signal cables in an orderly fashion. The pump and the optional column compartment each have one cable clip on each side and the autosampler has two cable clips on each side. The picture shows the location of the cable clips.
Figure: Cable clips in the system