Parts required
Digital I/O signaling cable
12
Figure: Digital I/O signaling cable
Pin
Description
1
6-pin connector, for connection to the Dig I/O port on the system
2
6 wires, for connection to an external instrument, for example, a mass spectrometer
Follow these steps
  1. On the autosampler, plug the 6-pin connector of the cable into the port labeled Dig I/O 2.
  2. Connect the wires for the output signals (white wire and brown wire) to the appropriate connectors on the mass spectrometer.
    With this, the inject response signal is delivered automatically.