To interrupt operation for a longer period and to keep the system in the laboratory, follow the instructions below.
Run the script: B05 - Shutdown (Long-term) with the following parameter: Module to remove = None.
TIP
Removing the solvent lines from the tubing guides is usually not required (not even for replacing the slide-in module). However, when you remove the solvent lines from the tubing guides in the pump and all modules above the pump in the system stack, be careful not to pull on other tubing in the guides.
Empty the solvent reservoirs.
Turn off the system with the system power button (lower button on the system base).
Turn off the system controller with its power button (upper button on the system base).
TIP
When the power is turned off to the autosampler, the left front door of the autosampler is opened automatically for proper ventilation of the sample compartment and cannot be closed.
Restarting the System after Long-Term Shutdown
Fill the solvent reservoirs. See Filling the Solvent Reservoirs.
Turn on the system. See Turning On the System for the First Time.
Prepare the system for operation. See Preparing the System for Operation.