When
After powering up the system and after preparing the system for initial operation
After a short-term shutdown
After changing the flow rate
After changing the temperature setpoints
Follow these steps
System equilibration should include the following:
Warming up (or cooling down) all temperature-controlled devices for a certain time:
Equilibration steps | Duration |
---|---|
Warming up (or cooling down) all temperature-controlled devices in the system to the starting temperature. Temperature-controlled devices can be, for example:
| 5 hours depending on the temperature differences needed for your application |
During warming up (or cooling down), do the following depending on the situation:
If you use the previously installed column: Equilibrate the column by running the script: B02 – Clean and Equilibrate Column(s).
If you installed a new, previously unused, column: To condition new columns, run the script: B06 – Condition Column(s).
Monitor the pump pressure drift and check that the drift is within reasonable limits for the application for 5-24 hours depending on the retention time accuracy needed for your application.