The rating plates are present on each system module near the electrical connections. The rating plate indicates the serial number, part number, module name, revision number, and the line and fuse rating.
System serial number label
In addition, a system serial number label is present at the inner side (bottom left) of the system base drawer. The system serial number label indicates the system serial number, part number, system name, and the revision number.
Type label
An additional type label on the leak tray of each system module indicates the module name, serial number, part number, and revision number. To facilitate device identification, have the information from this label available when communicating with Thermo Fisher Scientific.
System Controller
The rating plate is located on the short panel of the system controller. It indicates the serial number and the line rating.
Display
The rating plate is located on the rear side of the display. It indicates the module name, serial number, part number, revision number (if any), and the line rating.