When
Damage or blockage of waste line
Parts and additional items required
  • Flowmeter waste line
  • Solvent suitable to flush out harmful substances
Parts affected
12537416
Figure: Flowmeter waste line (left: overview; right: detailed view)
No.
Description
1
Flowmeter waste line
2
Seal wash detector assembly
3
Drain port
4
Installation position for seal wash detector assembly
5
Waste outlet port (to drain system)
6
Groove in drain port wall
7
Leak sensor
Preparations
  1. To flush out harmful substances, purge the pump with an appropriate solvent.
  2. For easier access to the drain port wall and waste outlet port, remove the seal wash detector assembly (funnel and detector).
Removing the Seal Wash Detector Assembly
Grasp and pull the seal wash detector assembly (funnel and detector) upward by the funnel. The detector including the tubing and cable can remain connected to the funnel.
Removing the seal wash detector assembly
Figure: Removing the seal wash detector assembly
No.
Description
1
Funnel, seal wash detector
2
Seal wash detector
3
Detector cable
4
Seal wash line
Follow these steps
  1. Remove the waste line from the Purge Out port on the flowmeter and from the waste outlet port.
  2. Connect the new waste line to the Purge Out port.
  3. Route the line through the groove in the drain port wall into the waste outlet port.
    To avoid false leak alarms, the waste line must sit properly in the waste outlet port. If it does not, liquid may accumulate in the drain port and activate the leak sensor.
  4. Reinsert the seal wash detector assembly.