When
Damage or blockage of the solvent line
Parts and additional items required
  • Solvent lines from shut-off valve to pump head
  • Solvent suitable to flush out harmful substances
  • Solvent suitable for your application
Preparations
  • To flush out harmful substances, purge the pump with an appropriate solvent.
Parts affected
432ABAB12
Figure: Solvent lines from shut-off valves to pump heads
No.
Description
1
Shut-off valves, A and B
2
Pump heads, A and B
3
Solvent line from shut-off valve B to pump head B
4
Solvent line from shut-off valve A to pump head A
Follow these steps
  1. Disconnect the solvent line on the shut-off valve and on the pump head.
  2. Install the new solvent line.
  3. Purge the pump using the solvent for your application.