When
During regular maintenance
Impaired permeability of solvent line filter frit
Damage of solvent line filter
Parts and additional items required
As applicable
Filter frit
Solvent line filter (with filter holder, filter frit, and solvent line adapter)
Solvent suitable for your application
Parts affected
No. | Description |
|---|---|
1 | Solvent line adapter |
2 | Filter holder, top part |
3 | Filter frit |
4 | Filter holder, bottom part |
Preparations
Unscrew the cap of the solvent reservoir and remove the cap together with the solvent line from the reservoir.
Remove the filter holder (with adapter) from the solvent line.
Follow these steps
As necessary, follow the steps for replacing the filter frit or for replacing the filter.
Steps for Replacing the Frit in the Solvent Line Filter
Open the filter holder and remove the filter frit.
Assemble the solvent line filter, wearing appropriate clean gloves:
- Place the frit in a level position in the filter holder (bottom part).
- Screw the filter top firmly to the filter bottom.
Slide the filter holder with the adapter onto the solvent line.
Retighten the reservoir cap hand-tight. The retaining guide must remain in the hole in the reservoir cap. If it does not, press the retaining guide into the hole so that the tubing is kept in place in the cap.
To remove any air from the solvent lines, purge the pump using the solvent of your application.
Steps for Replacing the Solvent Line Filter
Assemble the solvent line filter, wearing appropriate clean gloves:
- Insert the solvent line adapter into the filter holder (top part).
- Turn the filter holder (top part) with adapter upside down and press onto a suitable surface so that the adapter is seated firmly in the filter holder.
- Place the frit in a level position in the filter holder (bottom part).
- Screw the filter top firmly to the filter bottom.
Slide the filter holder with the adapter onto the solvent line.
Retighten the reservoir cap hand-tight. The retaining guide must remain in the hole in the reservoir cap. If it does not, press the retaining guide into the hole so that the tubing is kept in place in the cap.
Purge the pump using the solvent of your application.