The weight of the system and system modules are specified as follows:
Product | Weight | |
|---|---|---|
System | Basic configuration1: Approx. 66 kg | Basic configuration and column compartment: Approx. 79 kg |
Pump | 24 kg | |
Autosampler | 26 kg | |
Column compartment | Without valves: 10.7 kg; with 2 Vanquish valves: 11.6 kg; with 2 VICI valves: 13.1 kg | |
1: The basic configuration includes the system base with system controller, pump, autosampler and display | ||