The emission sound pressure levels are specified as follows:
Product | Emission sound pressure level |
|---|---|
System |
< 75 dB(A), typically < 55 dB(A) |
Pump | < 70 dB(A), typically < 50 dB(A) |
Autosampler | < 70 dB(A), typically < 50 dB(A) |
Column compartment | < 42 dB(A), typically < 40 dB(A) |