The dimensions of the system and system modules are specified as follows:
Product | Dimension (height x width x depth) | |
---|---|---|
System | Basic configuration1: 74 x 45 x 65 cm | Basic configuration and column compartment: 82 x 45 x 65 cm |
Pump | 19.2 x 42 x 62 cm | |
Autosampler | 29 x 42 x 62 cm | |
Column compartment | Outer dimension | 15.9 x 42 x 62 cm |
Column chamber | 7.8 x 34.3 x 3.7 cm | |
1: The basic configuration includes the system base with system controller, pump, autosampler and display |