The power requirements are specified as follows:
Product | Power requirements |
|---|---|
Pump | 100 – 240 V AC, ± 10 %; 50/60 Hz, max. 525 W / 550 VA |
Autosampler | 100 – 240 V AC, ± 10 %; 50/60 Hz; max. 525 W / 550 VA |
Column compartment | 100 – 240 V AC, ± 10 %; 50/60 Hz; max. 525 W / 550 VA |
System controller | 100 – 240 VAC, ± 10 %; 50/60 Hz, max. 50 W / 150 VA |