The environmental conditions are specified as follows:
Type | Specification |
---|---|
Range of use | Indoor use only |
Ambient operating temperature | 5 °C - 35 °C |
Ambient storage temperature | -20 °C - 45 °C |
Ambient operating humidity | 20% - 80% relative humidity (non-condensing) |
Ambient storage humidity | Maximum 60% relative humidity (non-condensing) |
Operating altitude | Maximum 2000 m above sea level |
Pollution degree | 2 |
Overvoltage category | II |